4-Chloro-2-fluoro-6-methylbenzoic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC535016-250MG
APOSPC535016-250MGEA 64.1 GBP
APOSPC535016-250MG
4-Chloro-2-fluoro-6-methylbenzoic acid 95%
4-Chloro-2-fluoro-6-methylbenzoic acid
Formula: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD23712076
CAS Number: 1427395-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR