5-Fluoro-2-iodoanisole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC53271-1G PC53271-5G
APOSPC53271-1GEA 87.3 GBP
APOSPC53271-1G APOSPC53271-5G
5-Fluoro-2-iodoanisole
5-Fluoro-2-iodoanisole
Formula: C₇H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD18396916
CAS Number: 450-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR