3-Fluoro-4-[4-(Boc-amino)piperidino]aniline 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC530054-1G PC530054-250MG PC530054-500MG
APOSPC530054-1GEA 118 GBP
APOSPC530054-1G APOSPC530054-250MG APOSPC530054-500MG
3-Fluoro-4-[4-(Boc-amino)piperidino]aniline 97%
3-Fluoro-4-[4-(Boc-amino)piperidino]aniline
Formula: C₁₆H₂₄FN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD23701381
CAS Number: 1000053-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR