2,5-Difluoro-D-phenylalanine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC530003-1G PC530003-250MG
APOSPC530003-1GEA 151 GBP
APOSPC530003-1G APOSPC530003-250MG
2,5-Difluoro-D-phenylalanine 97%
2,5-Difluoro-D-phenylalanine
Formula: C₉H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD07371999
CAS Number: 266360-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR