3-(Trifluoromethyl)-o-anisidine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 3-(Trifluoromethyl)-2-methoxyaniline, 3-(Trifluoromethyl)-2-anisidine, 2-Amino-6-(trifluoromethyl)anisole, 3-Amino-2-methoxybenzotrifluoride, 2-Methoxy-3-(trifluoromethyl)aniline

PC5268-10G PC5268-5G PC5268-1G PC5268-25G
APOSPC5268-10GEA 270 GBP
APOSPC5268-10G APOSPC5268-5G APOSPC5268-1G APOSPC5268-25G
3-(Trifluoromethyl)-o-anisidine 98%
3-(Trifluoromethyl)-o-anisidine
Formula: C₈H₈F₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD08532489
CAS Number: 634187-08-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR