3-Bromo-5-(trifluoromethoxy)benzal fluoride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC52181-1G PC52181-250MG PC52181-5G
APOSPC52181-1GEA 46.2 GBP
APOSPC52181-1G APOSPC52181-250MG APOSPC52181-5G
3-Bromo-5-(trifluoromethoxy)benzal fluoride
3-Bromo-5-(trifluoromethoxy)benzal fluoride
Formula: C₈H₄BrF₅O
Storage Temperature: Ambient
MDL Number: MFCD31812118
CAS Number: 2167884-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR