2,4-Dichloro-3-fluorobenzoic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC520904-5G
APOSPC520904-5GEA 343 GBP
APOSPC520904-5G
2,4-Dichloro-3-fluorobenzoic acid 95%
2,4-Dichloro-3-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD14707646
CAS Number: 915145-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR