Benzene-1,3-disulphonyl fluoride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC520783-1G PC520783-5G
APOSPC520783-1GEA 136 GBP
APOSPC520783-1G APOSPC520783-5G
Benzene-1,3-disulphonyl fluoride 95%
Benzene-1,3-disulphonyl fluoride
Formula: C₆H₄F₂O₄S₂
Melting Pt: 37…42 °C
Storage Temperature: Ambient
MDL Number: MFCD28346978
CAS Number: 7552-55-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR