4-[2-(Trifluoromethyl)phenoxy]piperidine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC520542-10G PC520542-1G PC520542-5G
APOSPC520542-10GEA 741 GBP
APOSPC520542-10G APOSPC520542-1G APOSPC520542-5G
4-[2-(Trifluoromethyl)phenoxy]piperidine 95%
4-[2-(Trifluoromethyl)phenoxy]piperidine
Formula: C₁₂H₁₄F₃NO
Storage Temperature: Ambient
MDL Number: MFCD06248881
CAS Number: 824390-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR