2-Chloro-6-(trifluoromethyl)isonicotinaldehyde

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC51496-1G PC51496-250MG
APOSPC51496-1GEA 436 GBP
APOSPC51496-1G APOSPC51496-250MG
2-Chloro-6-(trifluoromethyl)isonicotinaldehyde
2-Chloro-6-(trifluoromethyl)isonicotinaldehyde
Formula: C₇H₃ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD13188805
CAS Number: 1060807-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR