Methyl pentafluoromethacrylate 96%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC5128-5G PC5128-1G PC5128-250MG
APOSPC5128-5GEA 604 GBP
APOSPC5128-5G APOSPC5128-1G APOSPC5128-250MG
Methyl pentafluoromethacrylate 96%
Methyl pentafluoromethacrylate
Formula: C₅H₃F₅O₂
MW: 190.07 g/mol
Boiling Pt: 94…95 °C
Density: 1.373 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00153210
CAS Number: 685-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR