4-Chloro-6-(trifluoromethyl)nicotinic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-Chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid, 5-Carboxy-4-chloro-2-(trifluoromethyl)pyridine

PC51210-1G PC51210-250MG PC51210-5G
APOSPC51210-1GEA 267 GBP
APOSPC51210-1G APOSPC51210-250MG APOSPC51210-5G
4-Chloro-6-(trifluoromethyl)nicotinic acid
4-Chloro-6-(trifluoromethyl)nicotinic acid
Formula: C₇H₃ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD13188812
CAS Number: 1060810-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR