2-Chloro-5-fluoropyrimidin-4-one 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC51106-5G
APOSPC51106-5GEA 104 GBP
APOSPC51106-5G
2-Chloro-5-fluoropyrimidin-4-one 95+%
2-Chloro-5-fluoropyrimidin-4-one
Formula: C₄H₂ClFN₂O
Melting Pt: 176…177 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01346689
CAS Number: 155-12-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR