Ethyl 2-(4-Biphenylyl)-2,2-difluoroacetate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC510311-1G
APOSPC510311-1GEA 582 GBP
APOSPC510311-1G
Ethyl 2-(4-Biphenylyl)-2,2-difluoroacetate
Ethyl 2-(4-Biphenylyl)-2,2-difluoroacetate
Formula: C₁₆H₁₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26728234
CAS Number: 73789-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR