2-Nitro-4-(trifluoromethyl)anisole 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC5060-100G PC5060-10G PC5060-25G PC5060-5G
APOSPC5060-100GEA 104 GBP
APOSPC5060-100G APOSPC5060-10G APOSPC5060-25G APOSPC5060-5G
2-Nitro-4-(trifluoromethyl)anisole 97%
2-Nitro-4-(trifluoromethyl)anisole
Formula: C₈H₆F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD00007099
CAS Number: 394-25-2
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR