3-Nitro-5-(trifluoromethyl)anisole 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC5055-1G PC5055-25G PC5055-5G
APOSPC5055-1GEA 15.8 GBP
APOSPC5055-1G APOSPC5055-25G APOSPC5055-5G
3-Nitro-5-(trifluoromethyl)anisole 99%
3-Nitro-5-(trifluoromethyl)anisole
Formula: C₈H₆F₃NO₃
Melting Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00010446
CAS Number: 328-79-0
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR