4-Nitro-2-(trifluoromethyl)anisole 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC5050-100G PC5050-25G PC5050-500G
APOSPC5050-100GEA 93.2 GBP
APOSPC5050-100G APOSPC5050-25G APOSPC5050-500G
4-Nitro-2-(trifluoromethyl)anisole 99%
4-Nitro-2-(trifluoromethyl)anisole
Formula: C₈H₆F₃NO₃
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00007328
CAS Number: 654-76-2
EINECS: 211-507-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR