5-Bromo-2-(trifluoromethyl)benzyl fluoride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC502907-1G PC502907-250MG
APOSPC502907-1GEA 415 GBP
APOSPC502907-1G APOSPC502907-250MG
5-Bromo-2-(trifluoromethyl)benzyl fluoride 95%
5-Bromo-2-(trifluoromethyl)benzyl fluoride
Formula: C₈H₅BrF₄
Storage Temperature: Ambient
MDL Number: MFCD31634841
CAS Number: 2244086-36-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR