2-Bromo-3-fluorobenzyl fluoride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC502903-1G PC502903-250MG
APOSPC502903-1GEA 441 GBP
APOSPC502903-1G APOSPC502903-250MG
2-Bromo-3-fluorobenzyl fluoride 95%
2-Bromo-3-fluorobenzyl fluoride
Formula: C₇H₅BrF₂
Storage Temperature: Ambient
MDL Number: MFCD31634837
CAS Number: 2244088-41-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR