2-Bromo-6-fluorobenzyl fluoride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC502901-1G PC502901-250MG
APOSPC502901-1GEA 485 GBP
APOSPC502901-1G APOSPC502901-250MG
2-Bromo-6-fluorobenzyl fluoride 95%
2-Bromo-6-fluorobenzyl fluoride
Formula: C₇H₅BrF₂
MW: 207.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28527821
CAS Number: 1782526-83-3
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR