2-Bromo-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-(2-Bromo-4-methyl-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502766-1G
APOSPC502766-1GEA 848 GBP
APOSPC502766-1G
2-Bromo-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formula: C₁₄H₈BrF₅O
Storage Temperature: Ambient
MDL Number: MFCD31630071
CAS Number: 2244084-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR