2,6-Dichloro-4-fluorobenzoic acid ≥97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC502273-250MG PC502273-1G PC502273-5G
APOSPC502273-250MGEA 83.9 GBP
APOSPC502273-250MG APOSPC502273-1G APOSPC502273-5G
2,6-Dichloro-4-fluorobenzoic acid ≥97%
2,6-Dichloro-4-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₂
MDL Number: MFCD23135660
CAS Number: 232275-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR