Potassium [3-(trifluoromethyl)benzyl]trifluoroborate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC502198-1G PC502198-250MG PC502198-5G
APOSPC502198-1GEA 241 GBP
APOSPC502198-1G APOSPC502198-250MG APOSPC502198-5G
Potassium [3-(trifluoromethyl)benzyl]trifluoroborate 95%
Potassium [3-(trifluoromethyl)benzyl]trifluoroborate
Formula: C₈H₆BF₆·K
Storage Temperature: Ambient
MDL Number: MFCD18712965
CAS Number: 1494466-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR