2,4-Dichloro-7-fluoroquinazoline 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC502107-1G
APOSPC502107-1GEA 49.4 GBP
APOSPC502107-1G
2,4-Dichloro-7-fluoroquinazoline 95%
2,4-Dichloro-7-fluoroquinazoline
Formula: C₈H₃Cl₂FN₂
Storage Temperature: Ambient
MDL Number: MFCD09954885
CAS Number: 174566-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR