2,4-Dichloro-6-fluoroquinazoline 97+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC502106-1G PC502106-5G
APOSPC502106-1GEA 74.6 GBP
APOSPC502106-1G APOSPC502106-5G
2,4-Dichloro-6-fluoroquinazoline 97+%
2,4-Dichloro-6-fluoroquinazoline
Formula: C₈H₃Cl₂FN₂
MW: 217.0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09954884
CAS Number: 134517-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR