4-Bromo-3-(trifluoromethyl)phenacyl bromide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Bromo-5-(bromoacetyl)benzotrifluoride, 2-Bromo-1-[4-bromo-3-(trifluoromethyl)phenyl]ethan-1-one

PC500506-1G PC500506-250MG PC500506-5G
APOSPC500506-1GEA 294 GBP
APOSPC500506-1G APOSPC500506-250MG APOSPC500506-5G
4-Bromo-3-(trifluoromethyl)phenacyl bromide
4-Bromo-3-(trifluoromethyl)phenacyl bromide
Formula: C₉H₅Br₂F₃O
MW: 345.939 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22491116
CAS Number: 1523196-85-1
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR