3-(Bromomethyl)-5-fluorobenzoic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: α-Bromo-5-fluoro-m-toluic acid, 3-Carboxy-5-fluorobenzyl bromide

PC500474-1G PC500474-250MG PC500474-5G
APOSPC500474-1GEA 231 GBP
APOSPC500474-1G APOSPC500474-250MG APOSPC500474-5G
3-(Bromomethyl)-5-fluorobenzoic acid
3-(Bromomethyl)-5-fluorobenzoic acid
Formula: C₈H₆BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18378634
CAS Number: 1379351-68-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR