4-Bromo-3-fluoro-2-methoxyaniline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC500208-1G PC500208-5G PC500208-250MG
APOSPC500208-1GEA 209 GBP
APOSPC500208-1G APOSPC500208-5G APOSPC500208-250MG
4-Bromo-3-fluoro-2-methoxyaniline
4-Bromo-3-fluoro-2-methoxyaniline
Formula: C₇H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD22570403
CAS Number: 1137869-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR