5-Bromo-7-(trifluoromethyl)quinoxaline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC49608-1G PC49608-250MG PC49608-500MG PC49608-5G
APOSPC49608-1GEA 376 GBP
APOSPC49608-1G APOSPC49608-250MG APOSPC49608-500MG APOSPC49608-5G
5-Bromo-7-(trifluoromethyl)quinoxaline
5-Bromo-7-(trifluoromethyl)quinoxaline
Formula: C₉H₄BrF₃N₂
MW: 277.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18089243
CAS Number: 1287218-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR