3-Methoxy-5-(trifluoromethyl)benzyl alcohol 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC49338-10G PC49338-5G PC49338-1G
APOSPC49338-10GEA 294 GBP
APOSPC49338-10G APOSPC49338-5G APOSPC49338-1G
3-Methoxy-5-(trifluoromethyl)benzyl alcohol 97%
3-Methoxy-5-(trifluoromethyl)benzyl alcohol
Formula: C₉H₉F₃O₂
MW: 206.16 g/mol
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD09025413
CAS Number: 916420-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR