4-Bromo-3-fluorophenylacetylene 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Bromo-4-ethynyl-2-fluorobenzene

PC49282-1G PC49282-250MG PC49282-5G
APOSPC49282-1GEA 160 GBP
APOSPC49282-1G APOSPC49282-250MG APOSPC49282-5G
4-Bromo-3-fluorophenylacetylene 98%
4-Bromo-3-fluorophenylacetylene
Formula: C₈H₄BrF
MW: 199.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22460480
CAS Number: 1191063-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR