3,5-Difluoro-2-hydroxyphenacyl bromide 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC49202-1G PC49202-5G
APOSPC49202-1GEA 94.9 GBP
APOSPC49202-1G APOSPC49202-5G
3,5-Difluoro-2-hydroxyphenacyl bromide 95%
3,5-Difluoro-2-hydroxyphenacyl bromide
Formula: C₈H₅BrF₂O₂
Melting Pt: 67.1…71.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19053258
CAS Number: 1192815-24-9
UN: 3263
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR