Potassium trifluoro(prop-1-en-2-yl)borate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potassium Isopropenyltrifluoroborate

PC49166-5G PC49166-25G PC49166-100G
APOSPC49166-5GEA 15.8 GBP
APOSPC49166-5G APOSPC49166-25G APOSPC49166-100G
Potassium trifluoro(prop-1-en-2-yl)borate 95%
Potassium trifluoro(prop-1-en-2-yl)borate
Formula: C3H5BF3K
MW: 147.9763 g/mol
Melting Pt: 274…280 °C
Storage Temperature: Ambient
MDL Number: MFCD11052654
CAS Number: 395083-14-4
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR