2-Chloro-5-(trifluoromethyl)benzoic acid, Technical Grade

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Chloro-5-trifluoromethylbenzoic acid, 6-Chloro-α,α,α-trifluoro-m-toluic acid

PC48678-25G PC48678-5G
APOSPC48678-25GEA 21 GBP
APOSPC48678-25G APOSPC48678-5G
2-Chloro-5-(trifluoromethyl)benzoic acid, Technical Grade
2-Chloro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₄ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00061094
CAS Number: 657-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR