4-Nitro-3-(trifluoromethyl)phenol 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: α,α,α-Trifluoro-4-nitro-m-cresol, 5-Hydroxy-2-nitrobenzotrifluoride

PC4860-1G PC4860-25G PC4860-5G
APOSPC4860-1GEA 15.8 GBP
APOSPC4860-1G APOSPC4860-25G APOSPC4860-5G
4-Nitro-3-(trifluoromethyl)phenol 97%
4-Nitro-3-(trifluoromethyl)phenol
Formula: C₇H₄F₃NO₃
Boiling Pt: 135…138 °C/0.01mmHg
Melting Pt: 72…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00014714
CAS Number: 88-30-2
EINECS: 201-818-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR