(2S)-3,3,3-Trifluoropropane-1,2-diol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC48579-1G PC48579-250MG
APOSPC48579-1GEA 1210 GBP
APOSPC48579-1G APOSPC48579-250MG
(2S)-3,3,3-Trifluoropropane-1,2-diol
(2S)-3,3,3-Trifluoropropane-1,2-diol
Formula: C₃H₅F₃O₂
Boiling Pt: 87…88 °C (25 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD22199791
CAS Number: 148683-14-1
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR