4-(Trifluoromethyl)pyridazine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC48363-1G PC48363-500MG PC48363-5G
APOSPC48363-1GEA 190 GBP
APOSPC48363-1G APOSPC48363-500MG APOSPC48363-5G
4-(Trifluoromethyl)pyridazine 97%
4-(Trifluoromethyl)pyridazine
Formula: C₅H₃F₃N₂
Density: 1.4 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD11226564
CAS Number: 132537-44-1
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR