2-Chloro-4-(trifluoromethyl)benzoic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Chloro-4-trifluoromethylbenzoic acid, 2-Chloro-α,α,α-trifluoro-p-toluic acid

PC48310-25G PC48310-100G PC48310-5G
APOSPC48310-25GEA 65.1 GBP
APOSPC48310-25G APOSPC48310-100G APOSPC48310-5G
2-Chloro-4-(trifluoromethyl)benzoic acid 97%
2-Chloro-4-(trifluoromethyl)benzoic acid
Formula: C₈H₄ClF₃O₂
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD06208178
CAS Number: 23228-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR