HFPO oligomer acid fluorides (mixture) 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC4819-25G PC4819-5G
APOSPC4819-25GEA 253 GBP
APOSPC4819-25G APOSPC4819-5G
HFPO oligomer acid fluorides (mixture) 97%
HFPO oligomer acid fluorides (mixture)
Storage Temperature: Ambient
MDL Number: MFCD00163888
CAS Number: 65208-35-7
UN: 3265
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR