3,3,3-Trifluoropropylamine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 3-Amino-1,1,1-trifluoropropane

PC4603-1G PC4603-5G PC4603-500MG
APOSPC4603-1GEA 57.8 GBP
APOSPC4603-1G APOSPC4603-5G APOSPC4603-500MG
3,3,3-Trifluoropropylamine 97%
3,3,3-Trifluoropropylamine
Formula: C₃H₆F₃N
Boiling Pt: 67…68 °C
Storage Temperature: Ambient
MDL Number: MFCD01694854
CAS Number: 460-39-9
UN: 2924
ADR: 3+8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR