Ethyl 4-fluoropiperidine-4-carboxylate hydrochloride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC448012-1G PC448012-5G
APOSPC448012-1GEA 92.3 GBP
APOSPC448012-1G APOSPC448012-5G
Ethyl 4-fluoropiperidine-4-carboxylate hydrochloride 95%
Ethyl 4-fluoropiperidine-4-carboxylate hydrochloride
Formula: C₈H₁₄FNO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD09880187
CAS Number: 845909-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR