3-Fluoropyridine-2-carboxylic acid tert-butyl ester 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: tert-Butyl 3-fluoropicolinate

PC446144-1G
APOSPC446144-1GEA 49.4 GBP
APOSPC446144-1G
3-Fluoropyridine-2-carboxylic acid tert-butyl ester 95+%
3-Fluoropyridine-2-carboxylic acid tert-butyl ester
Formula: C₁₀H₁₂FNO₂
Storage Temperature: Ambient
MDL Number: MFCD29761614
CAS Number: 1934827-83-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR