2-Chloro-6-fluorophenylchlorooxime 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC446107-1G
APOSPC446107-1GEA 230 GBP
APOSPC446107-1G
2-Chloro-6-fluorophenylchlorooxime 95+%
2-Chloro-6-fluorophenylchlorooxime
Formula: C₇H₄Cl₂FNO
Storage Temperature: Ambient
MDL Number: MFCD24386151
CAS Number: 51088-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR