2-(1-(tert-Butoxycarbonyl)-4,4-difluoropyrrolidin-3-yl)acetic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC430227-1G
APOSPC430227-1GEA 1210 GBP
APOSPC430227-1G
2-(1-(tert-Butoxycarbonyl)-4,4-difluoropyrrolidin-3-yl)acetic acid
2-(1-(tert-Butoxycarbonyl)-4,4-difluoropyrrolidin-3-yl)acetic acid
Formula: C₁₁H₁₇F₂NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD27924137
CAS Number: 1780345-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR