Caesium perfluoroheptanoate 5 mM in water/acetonitrile (1:1)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC4289-10ML
APOSPC4289-10MLEA 62 GBP
APOSPC4289-10ML
Caesium perfluoroheptanoate 5 mM in water/acetonitrile (1:1)
Caesium perfluoroheptanoate
Formula: C₇F₁₃O₂·Cs
Storage Temperature: Ambient
MDL Number: MFCD04038276
CAS Number: 171198-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR