3-Chloro-2-fluorobenzoyl fluoride 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC4272-1G PC4272-250MG
APOSPC4272-1GEA 175 GBP
APOSPC4272-1G APOSPC4272-250MG
3-Chloro-2-fluorobenzoyl fluoride 98%
3-Chloro-2-fluorobenzoyl fluoride
Formula: C₇H₃ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD04972721
CAS Number: 85345-74-0
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR