3-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridin-3-ol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC421210-100MG
APOSPC421210-100MGEA 569 GBP
APOSPC421210-100MG
3-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridin-3-ol 95%
3-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridin-3-ol
Formula: C₈H₇F₃N₂O
MW: 204.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24556540
CAS Number: 892414-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR