Methyl 3,5-difluoro-4-formylbenzoate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC421099-250MG
APOSPC421099-250MGEA 54.6 GBP
APOSPC421099-250MG
Methyl 3,5-difluoro-4-formylbenzoate 97%
Methyl 3,5-difluoro-4-formylbenzoate
Formula: C₉H₆F₂O₃
MW: 200.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28525711
CAS Number: 1415124-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR