5-(Trifluoromethyl)-2-pyridineboronic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC412562-1G PC412562-250MG
APOSPC412562-1GEA 266 GBP
APOSPC412562-1G APOSPC412562-250MG
5-(Trifluoromethyl)-2-pyridineboronic acid
5-(Trifluoromethyl)-2-pyridineboronic acid
Formula: C₆H₅BF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD06227445
CAS Number: 1162257-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR