2-Ethyl-5-(trifluoromethyl)pyrazole-3-boronic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC412400-1G
APOSPC412400-1GEA 406 GBP
APOSPC412400-1G
2-Ethyl-5-(trifluoromethyl)pyrazole-3-boronic acid
2-Ethyl-5-(trifluoromethyl)pyrazole-3-boronic acid
Formula: C₆H₈BF₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD23135908
CAS Number: 1346665-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR